missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD56 (NCAM) Mouse anti-Human, PE-Cy5.5, Clone: CMSSB, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $402.60

Specifications

Antigen CD56 (NCAM)
Clone CMSSB
Host Species Murine
Gene Alias Neural Cell Adhesion Molecule
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123549 Life Technologies
LS35056741
25 Tests Each for $170.28
501123550 Life Technologies
LS35056742
100 Tests Each for $402.60
Specifications

Description

This CMSSB monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with CMSSB does not block binding of MEM188 or CB56.
Description & Specifications

Specifications

CD56 (NCAM)
Murine
Flow Cytometry
PE-Cy5.5
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.125μg)/test
CMSSB
Neural Cell Adhesion Molecule
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer