Please login to your online account to display your discounted pricing

CD56 (NCAM) Mouse anti-Human, PE, Clone: TULY56, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $346.55

Specifications

Antigen CD56 (NCAM)
Clone TULY56
Host Species Mouse
Gene Alias Neural cell adhesion molecule
Species Reactivity Human, Macaque, Monkey, Non-human primate, Rhesus Ma
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122239

 
life technologies
LS12056641
25 tests Each for $172.55

501122240

 
life technologies
LS12056642
100 tests Each for $346.55
Description & Specifications

Specifications

Antigen CD56 (NCAM)
Clone TULY56
Host Species Mouse
Gene Alias Neural cell adhesion molecule
Species Reactivity Human, Macaque, Monkey, Non-human primate, Rhesus Ma
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This TULY56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.

Staining with TULY56 does not block binding of CMSSB, suggesting that the two antibodies recognize different epitopes. Additionally, TULY56 performs better after fixation and permeabilization than CMSSB.

The TULY56 monoclonal antibody crossreacts with Rhesus macaque.