missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD56 (NCAM) Mouse anti-Human, FITC, Clone: TULY56, eBioscience™

Mouse Monoclonal Antibody

$147.90 - $295.80

Specifications

Antigen CD56 (NCAM)
Clone TULY56
Host Species Murine
Gene Alias Neural cell adhesion molecule
Species Reactivity Human, Macaque, Monkey, Non-human primate, Rhesus Ma
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122073 Life Technologies
LS11056642
100 Tests Each for $295.80
501129336 Life Technologies
LS11056641
25 Tests Each for $147.90
Specifications

Description

This TULY56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.

Staining with TULY56 does not block binding of CMSSB, suggesting that the two antibodies recognize different epitopes. Additionally, TULY56 performs better after fixation and permeabilization than CMSSB.

The TULY56 monoclonal antibody crossreacts with Rhesus macaque.
Description & Specifications

Specifications

CD56 (NCAM)
Murine
Human, Macaque, Monkey, Non-human primate, Rhesus Ma
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TULY56
Neural cell adhesion molecule
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test