Please login to your online account to display your discounted pricing

CD56 (NCAM) Mouse anti-Human, eFluor 660, Clone: 5tukon56, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $368.30

Specifications

Antigen CD56 (NCAM)
Clone 5tukon56
Host Species Murine
Gene Alias Neural cell adhesion molecule
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124273

 
life technologies
LS50056580
25μg Each for $146.45

501124274

 
life technologies
LS50056582
100μg Each for $368.30
Description & Specifications

Specifications

Antigen CD56 (NCAM)
Clone 5tukon56
Host Species Murine
Gene Alias Neural cell adhesion molecule
Species Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This 5tukon56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed in both the developing and adult nervous system and plays a role in neurogenesis, migration, neurite extension, and the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.

For flow cytometric analysis of human CD56, we recommend clones CMSSB or TULY56.