missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD56 (NCAM) Mouse anti-Human, Clone: CMSSB, eBioscience™

Mouse Monoclonal Antibody

$100.05 - $158.05

Specifications

Antigen CD56 (NCAM)
Clone CMSSB
Host Species Murine
Gene Alias Neural cell adhesion molecule
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012657 Life Technologies
LS14056780
25μg Each for $100.05
5012658 Life Technologies
LS14056782
100μg Each for $158.05
Specifications

Description

This CMSSB monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with CMSSB does not block binding of MEM188 or CB56.
Description & Specifications

Specifications

CD56 (NCAM)
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CMSSB
Neural cell adhesion molecule
Flow Cytometry,Immunohistochemistry (Formalin/Paraffin)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL