missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD56 (NCAM) Mouse anti-Human, Clone: 5tukon56, eBioscience™

Mouse Monoclonal Antibody

$92.40 - $146.52

Specifications

Antigen CD56 (NCAM)
Clone 5tukon56
Host Species Murine
Gene Alias Neural cell adhesion molecule
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122643 Life Technologies
LS14056580
25μg Each for $92.40
501122644 Life Technologies
LS14056582
100μg Each for $146.52
Specifications

Description

This 5tuckon56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed in both the developing and adult nervous system and plays a role in neurogenesis, migration, neurite extension, and the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.

For flow cytometric analysis of human CD56, we recommend clones CMSSB or TULY56.
Description & Specifications

Specifications

CD56 (NCAM)
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
5tukon56
Neural cell adhesion molecule
Immunohistochemistry (Formalin/Paraffin),Microscopy,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL