missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD56 (NCAM) Mouse anti-Human, APC-eFluor 780, Clone: CMSSB, eBioscience™

Mouse Monoclonal Antibody

$220.40 - $561.15

Specifications

Antigen CD56 (NCAM)
Clone CMSSB
Host Species Murine
Gene Alias Neural cell adhesion molecule
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245973 Life Technologies
LS47056741
25 Tests Each for $220.40
50245974 Life Technologies
LS47056742
100 Tests Each for $561.15
Specifications

Description

This CMSSB monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with CMSSB does not block binding of MEM188 or CB56.
Description & Specifications

Specifications

CD56 (NCAM)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CMSSB
Neural cell adhesion molecule
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.06μg)/test