Please login to your online account to display your discounted pricing

CD55 Mouse anti-Human, FITC, Clone: 143-30, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $288.55

Specifications

Antigen CD55
Clone 143-30
Host Species Mouse
Gene Alias Decay accelerating factor, DAF
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509622

 
affymetrix
11055941
25 tests Each for $143.55

509623

 
affymetrix
11055942
100 tests Each for $288.55
Description & Specifications

Specifications

Antigen CD55
Clone 143-30
Host Species Mouse
Gene Alias Decay accelerating factor, DAF
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 143-30 monoclonal antibody reacts with human CD55, the 64-73kDa molecule also known as DAF (Decay-Accelerating Factor). CD55 is a GPI-anchored single chain glycoprotein expressed on all cells in contact with serum including all hematopoietic cells and vascular endothelium; a soluble form of CD55 is also present in plasma and body fluids. CD55 is thought to play a role in the protection of cells from the damage by autologous complement via prevention of the amplification steps of the complement cascade. Antibodies against CD55 are mitogenic in the presence of phorbol esters.