Please login to your online account to display your discounted pricing

CD54 (ICAM-1) Rat anti-Mouse, FITC, Clone: YN1/1.7.4, eBioscience™

Rat Monoclonal Antibody

$114.55 - $404.55

Specifications

Antigen CD54 (ICAM-1)
Clone YN1/1.7.4
Host Species Rat
Gene Alias Intercellular adhesion molecule-1, ICAM1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122072

 
life technologies
LS11054181
50μg Each for $114.55

509620

 
life technologies
LS11054182
100μg Each for $216.05

509621

 
life technologies
LS11054185
500μg Each for $404.55
Description & Specifications

Specifications

Antigen CD54 (ICAM-1)
Clone YN1/1.7.4
Host Species Rat
Gene Alias Intercellular adhesion molecule-1, ICAM1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The YN1/1.7.4 monoclonal antibody reacts with mouse CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110kDa transmembrane glycoprotein expressed by monocytes, lymphocytes, dendritic cells, and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligands CD11a and CD11b is important in the inflammatory response. The YN1/1.7.4 monoclonal antibody recognizes a different epitope than the eBioKAT-1 monclonal antibody.