Please login to your online account to display your discounted pricing

CD54 (ICAM-1) Rat anti-Mouse, Clone: YN1/1.7.4, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$143.55 - $433.55

Specifications

Antigen CD54 (ICAM-1)
Clone YN1/1.7.4
Host Species Rat
Gene Alias Intercellular adhesion molecule-1, ICAM1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122943

 
life technologies
LS16054185
500μg Each for $433.55

5014346

 
life technologies
LS16054181
50μg Each for $143.55
Description & Specifications

Specifications

Antigen CD54 (ICAM-1)
Clone YN1/1.7.4
Host Species Rat
Gene Alias Intercellular adhesion molecule-1, ICAM1
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The YN1/1.7.4 monoclonal antibody reacts with mouse CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110kDa transmembrane glycoprotein expressed by monocytes, lymphocytes, dendritic cells, and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligands CD11a and CD11b is important in the inflammatory response. The YN1/1.7.4 monoclonal antibody recognizes a different epitope than the eBioKAT-1 monclonal antibody.