Please login to your online account to display your discounted pricing

CD54 (ICAM-1) Rat anti-Mouse, Clone: KAT-1, eBioscience™

Rat Monoclonal Antibody

$71.05 - $259.55

Specifications

Antigen CD54 (ICAM-1)
Clone KAT1)
Host Species Rat
Gene Alias Intercellular adhesion molecule-1, ICAM1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012650

 
life technologies
LS14054281
50μg Each for $71.05

5012651

 
life technologies
LS14054282
100μg Each for $143.55

5012652

 
life technologies
LS14054285
500μg Each for $259.55
Description & Specifications

Specifications

Antigen CD54 (ICAM-1)
Clone KAT1)
Host Species Rat
Gene Alias Intercellular adhesion molecule-1, ICAM1
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eBioKAT-1 monoclonal antibody reacts with mouse CD54 (ICAM-1), which is a 95kDa member of the immunoglobulin superfamily. CD54 is expressed at low levels on leukocytes and high endothelial venules, and expression increases in response to inflammatory cytokines. ICAM-1 binds to LFA-1 and this interaction is required for the transendothelial migration of T cells. ICAM-1-deficient mice are viable, however the migration of leukocytes to sites of inflammation is reduced leading to impaired immune and inflammatory responses. Based on the regulated expression of ICAM-1, it has been suggested that ICAM-1 may increase leukocyte extravasation at sites of inflammation, whereas the constitutively high expression of ICAM-2 mediates leukocyte traffic into non-inflamed tissue. The eBioKAT-1 monoclonal antibody recognizes a different epitope than the YN1/1.7.4 monoclonal antibody.