Please login to your online account to display your discounted pricing

CD54 (ICAM-1) Mouse anti-Human, Clone: HA58, eBioscience™

Mouse Monoclonal Antibody

$71.05 - $143.55

Specifications

Antigen CD54 (ICAM-1)
Clone HA58
Host Species Mouse
Gene Alias Intercellular adhesion molecule-1, ICAM1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012653

 
life technologies
LS14054980
25μg Each for $71.05

5012654

 
life technologies
LS14054982
100μg Each for $143.55
Description & Specifications

Specifications

Antigen CD54 (ICAM-1)
Clone HA58
Host Species Mouse
Gene Alias Intercellular adhesion molecule-1, ICAM1
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The HA58 monoclonal antibody reacts with human CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110kDa transmembrane glycoprotein expressed by monocytes, lymphocytes and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligand CD11a is important in the inflammatory response.