missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD54 (ICAM-1) Mouse anti-Human, APC, Clone: HA58, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $381.48

Specifications

Antigen CD54 (ICAM-1)
Clone HA58
Host Species Murine
Gene Alias Intercellular adhesion molecule-1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245650 Life Technologies
LS17054941
25 Tests Each for $157.08
50245651 Life Technologies
LS17054942
100 Tests Each for $381.48
Specifications

Description

The HA58 monoclonal antibody reacts with human CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110kDa transmembrane glycoprotein expressed by monocytes, lymphocytes and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligand CD11a is important in the inflammatory response.
Description & Specifications

Specifications

CD54 (ICAM-1)
Murine
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test
HA58
Intercellular adhesion molecule-1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer