Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD54/ICAM-1 Ab-4, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD54/ICAM-1 Ab-4, Mouse Monoclonal Antibody.

$274.31 - $1,133.78

Specifications

Antigen CD54/ICAM-1 Ab-4
Clone 23G12
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1094R7

 
thermo scientific™ lab vision™
MS1094R7
7mL Each for $961.94

MS1094S

 
thermo scientific™ lab vision™
MS1094S
1mL Each for $1,133.78

MS1094S0

 
thermo scientific™ lab vision™
MS1094S0
100µL Each for $274.31

MS1094S1

 
thermo scientific™ lab vision™
MS1094S1
500µL Each for $742.04
Description & Specifications

Specifications

Antigen CD54/ICAM-1 Ab-4
Clone 23G12
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Adhesion
Type Primary
Immunogen Recombinant protein encoding part of the external domain of human CD54
Isotype IgG1

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD54/ICAM-1 Ab-4, Mouse Monoclonal Antibody.

ICAM-1 (intercellular adhesion molecule) has 7 potential N-linked glycosylation sites. It is a single chain glycoprotein of Ig supergene family, present on unstimulated endothelial cells (EC) and on a variety of other cell types including activated fibroblasts, EC, macrophages, and lymphocytes. ICAM-1 mediates cell adhesion by binding to integrins CD11a/CD18 (leukocyte adhesion molecule, LFA-1) and to CD11b/CD18 (Mac-1).

Host Species: Mouse

Clone: 23G12

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant protein encoding part of the external domain of human CD54

Molecular Weight: 85-115 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting (Not suitable)

Note:

USA: RUO; Int'l: RUO