Please login to your online account to display your discounted pricing

Thermo Scientific™ CD54/ICAM-1 Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and functional study experiments.

$245.55 - $772.20

Specifications

Antigen CD54/ICAM-1 Ab-2
Clone W-CAM-1; same as Wehi-CAM-1 or 1H4
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS305P

 
thermo scientific™
MS305P
200μg/mL 1mL Each for $772.20

MS305P0

 
thermo scientific™
MS305P0
200μg/mL 100µL Each for $245.55

MS305P1

 
thermo scientific™
MS305P1
200μg/mL 500µL Each for $616.12

MS305P1ABX

 
thermo scientific™
MS305P1ABX
1mg/mL 100µL Each for $694.71
Description & Specifications

Specifications

Antigen CD54/ICAM-1 Ab-2
Clone W-CAM-1; same as Wehi-CAM-1 or 1H4
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Adhesion
Type Primary
Immunogen Raji Burkitt lymphoma cells
Isotype IgG2b kappa

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and functional study experiments with Thermo Scientific™ CD54/ICAM-1 Ab-2, Mouse Monoclonal Antibody.

ICAM-1 (intercellular adhesion molecule) has 7 potential N-linked glycosylation sites. It is a single chain glycoprotein of Ig supergene family, present on unstimulated endothelial cells (EC) and on a variety of other cell types including activated fibroblasts, EC, macrophages, and lymphocytes. ICAM-1 mediates cell adhesion by binding to integrins CD11a/CD18 (leukocyte adhesion molecule, LFA-1) and to CD11b/CD18 (Mac-1).

Host Species: Mouse

Clone: W-CAM-1; same as Wehi-CAM-1 or 1H4

Isotype: IgG2b/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Raji Burkitt lymphoma cells

Molecular Weight: 85-115kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Immunofluorescence
  • Flow Cytometry
  • Blocks intercellular adhesion of human cell lines
  • Functional Study

Note:

USA: RUO; Int'l: RUO