Please login to your online account to display your discounted pricing

Thermo Scientific™ CD54/ICAM-1 Ab-2, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and functional study experiments.

Manufacturer: thermo scientific™  MS305P1ABX

 View more versions of this product

Catalog No. MS305P1ABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2b kappa
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Quantity 100µL
Clone W-CAM-1; same as Wehi-CAM-1 or 1H4
Format Concentrated
Host Species Mouse
Immunogen Raji Burkitt lymphoma cells
Cross Reactivity Human
Antigen CD54/ICAM-1 Ab-2
Regulatory Status RUO
Research Discipline Cell Adhesion
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and functional study experiments with Thermo Scientific™ CD54/ICAM-1 Ab-2, Mouse Monoclonal Antibody.

ICAM-1 (intercellular adhesion molecule) has 7 potential N-linked glycosylation sites. It is a single chain glycoprotein of Ig supergene family, present on unstimulated endothelial cells (EC) and on a variety of other cell types including activated fibroblasts, EC, macrophages, and lymphocytes. ICAM-1 mediates cell adhesion by binding to integrins CD11a/CD18 (leukocyte adhesion molecule, LFA-1) and to CD11b/CD18 (Mac-1).

Host Species: Mouse

Clone: W-CAM-1; same as Wehi-CAM-1 or 1H4

Isotype: IgG2b/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Raji Burkitt lymphoma cells

Molecular Weight: 85-115kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Immunofluorescence
  • Flow Cytometry
  • Blocks intercellular adhesion of human cell lines
  • Functional Study

Note:

USA: RUO; Int'l: RUO