missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD54/ICAM-1 Ab-1, Mouse Monoclonal Antibody

Specifically detect human CD54 in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD54/ICAM-1 Ab-1, Mouse Monoclonal Antibody.

$254.41 - $861.10

Specifications

Antigen CD54/ICAM-1 Ab-1
Clone 15.2
Applications Flow Cytometry,Immunohistochemistry,Flow Cytometry,Immunohistochemistry
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS114P Thermo Scientific™ Lab Vision™
MS114P
200μg/mL 1mL Each for $765.78
MS114P0 Thermo Scientific™ Lab Vision™
MS114P0
200μg/mL 100μL Each for $254.41
MS114P1ABX Thermo Scientific™ Lab Vision™
MS114P1ABX
1mg/mL 100μL Each for $686.45
MS114PABX Thermo Scientific™ Lab Vision™
MS114PABX
1mg/mL 200μL Each for $861.10
Specifications

Description

Specifically detect human CD54 in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD54/ICAM-1 Ab-1, Mouse Monoclonal Antibody.

ICAM-1 (intercellular adhesion molecule) has 7 potential N-linked glycosylation sites. It is a single chain glycoprotein of Ig supergene family, present on unstimulated endothelial cells (EC) and on a variety of other cell types including activated fibroblasts, EC, macrophages, and lymphocytes. ICAM-1 mediates cell adhesion by binding to integrins CD11a/CD18 (leukocyte adhesion molecule, LFA-1) and to CD11b/CD18 (Mac-1). This interaction enhances antigen-specific T-cell activation. ICAM-1 also binds to CD43 and to Plasmodium falciparum infected RBCs.

Host Species: Mouse

Clone: 15.2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human monocytes

Molecular Weight: 85-115 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Acetone Fixed)
  • Flow Cytometry
  • Blocks binding of LFA-1 and infected RBCs to ICAM-3 (Order Ab without azide)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

CD54/ICAM-1 Ab-1
Flow Cytometry,Immunohistochemistry,Flow Cytometry,Immunohistochemistry
Unlabeled
Murine
Cell Adhesion
Human monocytes
15.2
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa