Please login to your online account to display your discounted pricing

CD51 (Integrin alpha V) Rat anti-Mouse, Biotin, Clone: RMV-7, eBioscience™

Rat Monoclonal Antibody

$100.05 - $361.05

Specifications

Antigen CD51 (Integrin alpha V)
Clone RMV-7
Host Species Rat
Gene Alias Integrin aV, ITGAV, Vitronectin receptor
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011585

 
life technologies
LS13051281
50μg Each for $100.05

5011586

 
life technologies
LS13051282
100μg Each for $172.55

5011587

 
life technologies
LS13051285
500μg Each for $361.05
Description & Specifications

Specifications

Antigen CD51 (Integrin alpha V)
Clone RMV-7
Host Species Rat
Gene Alias Integrin aV, ITGAV, Vitronectin receptor
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The RMV-7 monoclonal antibody reacts with the mouse CD51 molecule, the integrin alpha v chain. This approximately 120kDa surface molecule non-covalently associates with the β subunits of the integrin family including β3 (CD61), β1 (CD29), β5 and β6 to form receptors for extracellular matrix components. Heterodimers of CD51/CD61 are expressed by platelets, T cells and granulocytes and mediate adhesion to fibrinogen, fibronectin, vitronectin and thrombospondin.