missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD51/CD61 (Integrin alpha v beta 3) Mouse anti-Human, Clone: 23C6, eBioscience™

Mouse Monoclonal Antibody

$64.68 - $130.68

Specifications

Antigen CD51/CD61 (Integrin alpha v beta 3)
Clone 23C6
Host Species Murine
Gene Alias Integrin av b3, vitronectin Receptor
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012645 Life Technologies
LS14051980
25μg Each for $64.68
5012646 Life Technologies
LS14051982
100μg Each for $130.68
Specifications

Description

The 23C6 monoclonal antibody reacts with the human CD51/CD61 dimer, also known as the integrin αv/β3. CD51, an ∽120kDa surface molecule can also non-covalently associate with other β subunits of the integrin family including β1 (CD29), β5 and β6 to form receptors for extracellular matrix components. Heterodimers of CD51/CD61 are expressed by melanoma cells, endothelial cells and osteoclasts and at very low levels by platelets. The CD51/CD61 complex mediates adhesion to fibrinogen, fibronectin, vitronectin and thrombospondin.
Description & Specifications

Specifications

CD51/CD61 (Integrin alpha v beta 3)
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
23C6
Integrin av b3, vitronectin Receptor
Flow Cytometry,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL