Please login to your online account to display your discounted pricing

CD51/CD61 (Integrin alpha v beta 3) Mouse anti-Human, Biotin, Clone: 23C6, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $274.05

Specifications

Antigen CD51/CD61 (Integrin alpha v beta 3)
Clone 23C6
Host Species Mouse
Gene Alias Integrin av b3, vitronectin Receptor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011588

 
affymetrix
13051980
25μg Each for $114.55

5011589

 
affymetrix
13051982
100μg Each for $274.05
Description & Specifications

Specifications

Antigen CD51/CD61 (Integrin alpha v beta 3)
Clone 23C6
Host Species Mouse
Gene Alias Integrin av b3, vitronectin Receptor
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 23C6 monoclonal antibody reacts with the human CD51/CD61 dimer, also known as the integrin αv/β3. CD51, an ∽120kDa surface molecule can also non-covalently associate with other β subunits of the integrin family including β1 (CD29), β5 and β6 to form receptors for extracellular matrix components. Heterodimers of CD51/CD61 are expressed by melanoma cells, endothelial cells and osteoclasts and at very low levels by platelets. The CD51/CD61 complex mediates adhesion to fibrinogen, fibronectin, vitronectin and thrombospondin.