Please login to your online account to display your discounted pricing

CD5 Rat anti-Mouse, PE, Clone: 53-7.3, eBioscience™

Rat Monoclonal Antibody

$114.55 - $404.55

Specifications

Antigen CD5
Clone 53-7.3
Host Species Rat
Gene Alias Ly-1, Ly-12, Ly-A, Lyt-1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010173

 
life technologies
LS12005181
50μg Each for $114.55

5010176

 
life technologies
LS12005183
200μg Each for $404.55

501122206

 
life technologies
LS12005182
100μg Each for $216.05
Description & Specifications

Specifications

Antigen CD5
Clone 53-7.3
Host Species Rat
Gene Alias Ly-1, Ly-12, Ly-A, Lyt-1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 53-7.3 monoclonal antibody reacts with mouse CD5, a 67kDa protein expressed by a majority of thymocytes, mature T cells and a subset of B cells. The expression of CD5 by a small subset of B cells characterizes a developmentally and functionally distinct lineage of B cells called B-1 cells. CD5 is a counter-receptor for CD72 and plays a role in the T-B cell interaction.