missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD5 Rat anti-Mouse, FITC, Clone: 53-7.3, eBioscience™

Rat Monoclonal Antibody

$71.05 - $375.55

Specifications

Antigen CD5
Clone 53-7.3
Host Species Rat
Gene Alias Ly-1, Ly-12, Ly-A, Lyt-1
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122030 Life Technologies
LS11005182
100μg Each for $187.05
509397 Life Technologies
LS11005181
50μg Each for $71.05
509399 Life Technologies
LS11005185
500μg Each for $375.55
Specifications

Description

The 53-7.3 monoclonal antibody reacts with mouse CD5, a 67kDa protein expressed by a majority of thymocytes, mature T cells and a subset of B cells. The expression of CD5 by a small subset of B cells characterizes a developmentally and functionally distinct lineage of B cells called B-1 cells. CD5 is a counter-receptor for CD72 and plays a role in the T-B cell interaction.
Description & Specifications

Specifications

CD5
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
53-7.3
Ly-1, Ly-12, Ly-A, Lyt-1
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL