Please login to your online account to display your discounted pricing

CD5 Rat anti-Mouse, Biotin, Clone: 53-7.3, eBioscience™

Rat Monoclonal Antibody

$85.55 - $361.05

Specifications

Antigen CD5
Clone 53-7.3
Host Species Rat
Gene Alias Ly-1, Ly-12, Ly-A, Lyt-1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122501

 
affymetrix
13005181
50μg Each for $85.55

5011401

 
affymetrix
13005182
100μg Each for $143.55

5011402

 
affymetrix
13005185
500μg Each for $361.05
Description & Specifications

Specifications

Antigen CD5
Clone 53-7.3
Host Species Rat
Gene Alias Ly-1, Ly-12, Ly-A, Lyt-1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 53-7.3 monoclonal antibody reacts with mouse CD5, a 67kDa protein expressed by a majority of thymocytes, mature T cells and a subset of B cells. The expression of CD5 by a small subset of B cells characterizes a developmentally and functionally distinct lineage of B cells called B-1 cells. CD5 is a counter-receptor for CD72 and plays a role in the T-B cell interaction.