Please login to your online account to display your discounted pricing

CD5 Rat anti-Canine, PerCP-eFluor 710, Clone: YKIX322.3, eBioscience™

Rat Monoclonal Antibody

$143.55 - $368.30

Specifications

Antigen CD5
Clone YKIX322.3
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016057

 
life technologies
LS46505041
25 Tests Each for $143.55

5016058

 
life technologies
LS46505042
100 Tests Each for $368.30
Description & Specifications

Specifications

Antigen CD5
Clone YKIX322.3
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This YKIX322.3 monoclonal antibody reacts with canine CD5, also known as Ly-1. CD5 is a 67kDa type I transmembrane glycoprotein expressed on thymocytes and mature T cells. Studies have demonstrated that NK cells in dogs also express CD5, but at lower levels. In dogs, CD5 is not detectable on B cells or B cell lines.