Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD5, Rabbit Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry and western blotting procedures with Thermo Scientific™ CD5, Rabbit Monoclonal Antibody. 

$301.33 - $1,347.33

Specifications

Antigen CD5
Clone SP19
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RM9119R7

 
thermo scientific™ lab vision™
RM9119R7
7mL Each for $685.35

RM9119S

 
thermo scientific™ lab vision™
RM9119S
1mL Each for $1,347.33

RM9119S0

 
thermo scientific
RM9119S0
100µL Each for $301.33

RM9119S1

 
thermo scientific™ lab vision™
RM9119S1
500µL Each for $1,034.45
Description & Specifications

Specifications

Antigen CD5
Clone SP19
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status IVD
Research Discipline Hematopoietic
Type Primary
Immunogen Synthetic peptide from the intracellular region of CD5

Obtain accurate and reproducible results in immunohistochemistry and western blotting procedures with Thermo Scientific™ CD5, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

CD5, a transmembrane protein, is found on 95%of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.

Host Species: Rabbit

Clone: SP19

Species Reactivity: Human. Others not tested.

Epitope: Intracellular region

Immunogen: Synthetic peptide from the intracellular region of CD5

Positive Control: Raji cells, Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting