Please login to your online account to display your discounted pricing

CD5 Mouse anti-Human, PE-Cy7, Clone: UCHT2, eBioscience™

Mouse Monoclonal Antibody

$234.90 - $442.25

Specifications

Antigen CD5
Clone UCHT2
Host Species Mouse
Gene Alias Leu-1, T1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015443

 
life technologies
LS25005941
25 tests Each for $234.90

5015444

 
life technologies
LS25005942
100 tests Each for $442.25
Description & Specifications

Specifications

Antigen CD5
Clone UCHT2
Host Species Mouse
Gene Alias Leu-1, T1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The UCHT2 monoclonal antibody reacts with human CD5, a 67kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation.\\\\The monoclonal antibody UCHT2 recognizes primate CD5.