missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD5 Mouse anti-Human, PE-Cy5, Clone: L17F12, eBioscience™

Mouse Monoclonal Antibody

$183.48 - $447.48

Specifications

Antigen CD5
Clone L17F12
Host Species Murine
Gene Alias Leu-1, T1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122903 Life Technologies
LS15005841
25 Tests Each for $183.48
501122904 Life Technologies
LS15005842
100 Tests Each for $447.48
Specifications

Description

The L17F12 monoclonal antibody reacts with human CD5, a 67kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation.
Description & Specifications

Specifications

CD5
Murine
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test
L17F12
Leu-1, T1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer