missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD5 Mouse anti-Human, FITC, Clone: L17F12, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $240.24

Specifications

Antigen CD5
Clone L17F12
Host Species Murine
Gene Alias Leu-1, T1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509400 Life Technologies
LS11005841
25 Tests Each for $104.28
509401 Life Technologies
LS11005842
100 Tests Each for $240.24
Specifications

Description

The L17F12 monoclonal antibody reacts with human CD5, a 67kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation.
Description & Specifications

Specifications

CD5
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
L17F12
Leu-1, T1
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test