Please login to your online account to display your discounted pricing

CD49f (Integrin alpha 6) Rat anti-Human, Mouse, PerCP-eFluor 710, Clone: eBioGoH3 (GoH3), eBioscience™

Rat Monoclonal Antibody

$230.55 - $230.55

Specifications

Antigen CD49f (Integrin alpha 6)
Clone eBioGoH3 (GoH3)
Host Species Rat
Gene Alias Integrin a6, ITGA6, VLA6
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015973

 
life technologies
LS46049580
25μg Each for $230.55
Description & Specifications

Specifications

Antigen CD49f (Integrin alpha 6)
Clone eBioGoH3 (GoH3)
Host Species Rat
Gene Alias Integrin a6, ITGA6, VLA6
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBioGoH3 monoclonal antibody reacts with mouse and human CD49f, also known as integrin alpha 6, very late activation antigen 6 (VLA-6 alpha chain), and platelet gpIc. CD49f is a 120 kD transmembrane protein. CD49f associates with CD29, the integrin beta 1 chain, to form the VLA-6 complex; CD49f also associates with CD104, the integrin beta 4 chain, to form the alpha 6 beta 4 complex. CD49f is expressed primarily on T cells, monocytes, platelets, epithelial and endothelial cells. CD49f expression has also been found on germinal center B cells. The eBioGoH3 antibody is cross-reactive to integrin alpha 6 on human, mouse and bovine cells. This antibody has also been reported to have functional activity in blocking the binding of integrin alpha 6 to laminin.