missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD49f (Integrin alpha 6) Rat anti-Human, Mouse, PE-Cy7, Clone: eBioGoH3 (GoH3), eBioscience™

Rat Monoclonal Antibody

$200.64 - $443.52

Specifications

Antigen CD49f (Integrin alpha 6)
Clone eBioGoH3 (GoH3)
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245727 Life Technologies
LS25049580
25μg Each for $200.64
50245728 Life Technologies
LS25049582
100μg Each for $443.52
Specifications

Description

The eBioGoH3 monoclonal antibody reacts with mouse and human CD49f, also known as integrin alpha 6, very late activation antigen 6 (VLA-6 alpha chain), and platelet gpIc. CD49f is a 120 kD transmembrane protein. CD49f associates with CD29, the integrin beta 1 chain, to form the VLA-6 complex; CD49f also associates with CD104, the integrin beta 4 chain, to form the alpha 6 beta 4 complex. CD49f is expressed primarily on T cells, monocytes, platelets, epithelial and endothelial cells. CD49f expression has also been found on germinal center B cells. The eBioGoH3 antibody is cross-reactive to integrin alpha 6 on human, mouse and bovine cells. This antibody has also been reported to have functional activity in blocking the binding of integrin alpha 6 to laminin.
Description & Specifications

Specifications

CD49f (Integrin alpha 6)
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioGoH3 (GoH3)
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL