missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD49e (Integrin alpha 5) Mouse anti-Human, PE, Clone: P1D6, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $328.68

Specifications

Antigen CD49e (Integrin alpha 5)
Clone P1D6
Host Species Murine
Gene Alias ITGA5, VLA-5, Fibronectin receptor
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010797 Life Technologies
LS12490041
25 Tests Each for $157.08
501122317 Life Technologies
LS12490042
100 Tests Each for $328.68
Specifications

Description

This P1D6 monoclonal antibody reacts with human CD49e, which is also known as integrin alpha 5. Integrins constitute a family of cell surface receptors that mediate adhesion between cells or between a cell and the extracellular matrix. These proteins are heterodimers consisting of two distinct subunits, α and β. Integrin alpha 5 is expressed on thymocytes, T cells, monocytes, platelets, as well as early and activated B cells. Integrins are also expressed on non-hematopoietic and cancer cells. Integrin-mediated adhesion is regulated by changes in ligand affinity (or activation), cell shape, and integrin clustering and diffusion. Integrin alpha 5 undergoes post-translational cleavage in its extracellular domain to produce disulfide-linked light and heavy chains that associate with the common beta 1 subunit to form the fibronectin receptor. Studies have demonstrated the involvement of this integrin in cell adhesion, signaling, and apoptosis.

The P1D6 monoclonal antibody has been reported to have blocking activity. Crossblocking studies indicate that P1D6 and eBioSAM-1 recognize a similar epitope.
Description & Specifications

Specifications

CD49e (Integrin alpha 5)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
P1D6
ITGA5, VLA-5, Fibronectin receptor
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer