missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD49e (Integrin alpha 5) Mouse anti-Human, Alexa Fluor 488, Clone: SAM-1, eBioscience™

Mouse Monoclonal Antibody

$196.68 - $394.68

Specifications

Antigen CD49e (Integrin alpha 5)
Clone SAM1)
Host Species Murine
Gene Alias Integrin a5, ITGA5, VLA5A
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016704 Life Technologies
LS53049641
25 Tests Each for $196.68
5016705 Life Technologies
LS53049642
100 Tests Each for $394.68
Specifications

Description

The eBioSAM-1 monoclonal antibody reacts with human integrin alpha 5, also known as fibronectin receptor alpha chain, very late activation antigen 5 alpha, and CD49e. Integrins are composed of an alpha chain and a beta chain, which non-covalently associate to form the functional integrin. Integrin heterodimers participate in cell surface-mediated signaling and adhesion functions. Integrin alpha 5 undergoes post-translational cleavage in its extracellular domain to yield disulfide linked light and heavy chains that join with Integrin beta 1 (CD29) to form the fibronectin receptor, also known as the very late activation antigen-5 (VLA-5) complex. Integrin alpha 5 is expressed on thymocytes, T cells, monocytes, platelets, early B cells, and activated B cells.
Description & Specifications

Specifications

CD49e (Integrin alpha 5)
Murine
Flow Cytometry
Alexa Fluor 488
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
SAM1)
Integrin a5, ITGA5, VLA5A
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer