missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD49d (Integrin alpha 4) Mouse anti-Human, Clone: 9F10, eBioscience™

Mouse Monoclonal Antibody

$64.68 - $130.68

Specifications

Antigen CD49d (Integrin alpha 4)
Clone 9F10
Host Species Murine
Gene Alias Integrin a4, VLA4, ITGA4
Applications Flow Cytometry,Functional Assay,Immunohistochemistry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012640 Life Technologies
LS14049980
25μg Each for $64.68
5012641 Life Technologies
LS14049982
100μg Each for $130.68
Specifications

Description

The 9F10 monoclonal antibody reacts with human CD49d, the 150kDa integrin α4 subunit. The complex of CD49d non-covalently associated with integrin β1 (CD29), also known as VLA-4, is a receptor for fibronectin and VCAM-1 (CD106). This complex is expressed by thymocytes, peripheral lymphocytes, monocytes and eosinophils. CD49d also associates with integrin β7 and binds to the Mucosal Addressin Cell-Adhesion Molecule-1 (MadCAM-1).
Description & Specifications

Specifications

CD49d (Integrin alpha 4)
Murine
Flow Cytometry,Functional Assay,Immunohistochemistry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
9F10
Integrin a4, VLA4, ITGA4
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer