missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD49b (Integrin alpha 2) Rat, eFluor 506, Clone: DX5, eBioscience™

Rat Monoclonal Antibody

$130.68 - $381.48

Specifications

Antigen CD49b (Integrin alpha 2)
Clone DX5
Host Species Rat
Gene Alias DX5, pan-NK
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124979 Life Technologies
LS69597180
25μg Each for $130.68
501124980 Life Technologies
LS69597182
100μg Each for $381.48
Specifications

Description

The DX5 monoclonal antibody reacts with CD49b, an antigen expressed on a majority of mouse natural killer cells and a subset of T cells. DX5 reacts with all strains of mouse tested including the most commonly used strains, BALB/c, C57BL/6, C3H, CBA, DBA, AKR, SJL and 129. Simultaneous staining of C57BL/6 spleen cells with anti-NK1.1 mAb (PK136) and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.
Description & Specifications

Specifications

CD49b (Integrin alpha 2)
Rat
Flow Cytometry
eFluor 506
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
DX5
DX5, pan-NK
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer