missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD49b (Integrin alpha 2), eFluor 506, clone: DX5, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS69597180

 View more versions of this product

Catalog No. 501124979



Description

Description

GTP-binding proteins (G-proteins) play a critical role in signal transduction by coupling receptors to effectors. The intracellular consequences of G-protein activation include second messenger generation, protein phosphorylation, ion channel activation, gene induction, cell growth, and differentiation. Gi belongs to the pertussis toxin sensitive G-proteins. Gi consists of three different subtypes of a-subunits (ai1, ai2 and ai3).

Description: The DX5 monoclonal antibody reacts with CD49b, an antigen expressed on a majority of mouse natural killer cells and a subset of T cells. DX5 reacts with all strains of mouse tested including the most commonly used strains, BALB/c, C57BL/6, C3H, CBA, DBA, AKR, SJL and 129. Simultaneous staining of C57BL/6 spleen cells with anti-NK1.1 mAb (PK136) and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.Applications Reported: This DX5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This DX5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor™ 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD49b (Integrin alpha 2)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q62469
BR, CD49B, GPIa, VLA-2, VLAA2, ITGA2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
DX5
eFluor 506
Itga2
Liquid
Itga2
IgM, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16398
Documents

For Research Use Only.