Please login to your online account to display your discounted pricing

CD49b (Integrin alpha 2) Rat anti-Mouse, PE-eFluor 610, Clone: DX5, eBioscience™

Rat Monoclonal Antibody

$158.05 - $382.80

Specifications

Antigen CD49b (Integrin alpha 2)
Clone DX5
Host Species Rat
Gene Alias DX5, pan-NK
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124858

 
life technologies
LS61597180
25μg Each for $158.05

501124859

 
life technologies
LS61597182
100μg Each for $382.80
Description & Specifications

Specifications

Antigen CD49b (Integrin alpha 2)
Clone DX5
Host Species Rat
Gene Alias DX5, pan-NK
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The DX5 monoclonal antibody reacts with CD49b, an antigen expressed on a majority of mouse natural killer cells and a subset of T cells. DX5 reacts with all strains of mouse tested including the most commonly used strains, BALB/c, C57BL/6, C3H, CBA, DBA, AKR, SJL and 129. Simultaneous staining of C57BL/6 spleen cells with anti-NK1.1 mAb (PK136) and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.