Please login to your online account to display your discounted pricing

CD49b (Integrin alpha 2) Armenian Hamster anti-Mouse, Clone: HMa2, Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody, Functional Grade

$143.55 - $433.55

Specifications

Antigen CD49b (Integrin alpha 2)
Clone HMa2
Host Species Armenian Hamster
Gene Alias Integrin a2, VLA2, ITGA2, DX5
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014333

 
life technologies
LS16049181
50μg Each for $143.55

5014334

 
life technologies
LS16049185
500μg Each for $433.55
Description & Specifications

Specifications

Antigen CD49b (Integrin alpha 2)
Clone HMa2
Host Species Armenian Hamster
Gene Alias Integrin a2, VLA2, ITGA2, DX5
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The HMα2 monoclonal antibody reacts with mouse CD49b, the 150kDa integrin α2 subunit. The complex of CD49b non-covalently associated with integrin β1 (CD29), also known as VLA-2, is a receptor for collagen and laminin. This complex is expressed by some CD4+ T cells, IEL, NK cells, platelets and epithelial cells.