missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD48 Mouse anti-Human, Clone: eBio156-4H9 (156-4H9), Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$130.68 - $473.88

Specifications

Antigen CD48
Clone eBio156-4H9 (156-4H9)
Host Species Murine
Gene Alias BCM1, BCM 1, B-cell membrane protein, SLAMF2, SLAM F2, BLAST1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014331 Life Technologies
LS16048981
50μg Each for $130.68
5014332 Life Technologies
LS16048985
500μg Each for $473.88
Specifications

Description

The eBio156-4H9 monoclonal antibody reacts with the human CD48 antigen, also known as B cell activation marker 1, SLAM family member 2, Blast-1 (human), and OX-45 (rat). CD48, a member of the SLAM family and Ig superfamily, is a 45kDa GPI-linked glycoprotein expressed on the majority of hematopoietic cells. CD2 is the low affinity ligand for CD48.Recent publications have reported differential expression in mice of members of the SLAM family including CD48, CD150, and CD244 among functionally distinct bone marrow hematopoietic progenitors providing a useful tool for prediction of the primitiveness of hematopoietic progenitors. Expression of CD48 in hematopoietic cells is found on many leukocytes with little to none on granulocytes and absent on platelets and erythrocytes. Expression increases on B cells with activation.
Description & Specifications

Specifications

CD48
Murine
Human
RUO
Purified
Primary
aqueous buffer, no sodium azide
eBio156-4H9 (156-4H9)
BCM1, BCM 1, B-cell membrane protein, SLAMF2, SLAM F2, BLAST1
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL