missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD47 Rat anti-Mouse, Clone: miap301, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$143.55 - $520.55

Specifications

Antigen CD47
Clone miap301
Host Species Rat
Gene Alias Integrin associated protein, IAP
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014324 Life Technologies
LS16047181
50μg Each for $143.55
5014325 Life Technologies
LS16047185
500μg Each for $520.55
Specifications

Description

The monoclonal antibody miap301 reacts to CD47 also known as integrin-associated protein (IAP), and neurophilin. CD47 is a glycosylated five transmembrane protein with a small cytoplasmic domain. CD47 is involved in adhesion through interactions with SIRP (signal regulator protein) and is non-covalently associated with ß3 integrins CD51/CD61, CD41/CD61. It is also known to be the receptor for thrombospondin. T cell expression of CD47 can mediate activation or apoptosis (in the presence of high levels of thrombospondin). Expression is found in the majority of hematopoietic cells including T and B cells, monocytes, platelets and erythrocytes (as part of the Rh complex). Expression is also found in non-hematopoietic cells.

No crossreactivity is seen against human CD47.
Description & Specifications

Specifications

CD47
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, no sodium azide
miap301
Integrin associated protein, IAP
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL