missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD47 Mouse anti-Human, FITC, Clone: 2D3, eBioscience™

Mouse Monoclonal Antibody

$130.50 - $307.40

Specifications

Antigen CD47
Clone 2D3
Host Species Murine
Gene Alias Integrin associated protein, IAP
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122069 Life Technologies
LS11047841
25 Tests Each for $130.50
501129192 Life Technologies
LS11047842
100 Tests Each for $307.40
Specifications

Description

The monoclonal antibody 2D3 reacts to CD47 also known as integrin-associated protein (IAP), and neurophilin. CD47 is a glycosylated five transmembrane protein with a small alternatively spliced cytoplasmic domain. CD47 is involved in adhesion through interactions with SIRP (signal regulator protein) and is non-covalently associated with β3 integrins CD51/CD61 and CD41/CD61. Furthermore this interaction can mediate bi-directional signaling to modify neural synaptic activity and regulate the phagocytic activities of macrophages. CD47 is the receptor for thrombospondin. T cell expression of CD47 can mediate activation or apoptosis (in the presence of high levels of thrombospondin). Expression is found in the majority of hematopoietic cells including T and B cells, monocytes, platelets and erythrocytes (as part of the Rh complex). Expression is also found in non-hematopoietic cells.
Description & Specifications

Specifications

CD47
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2D3
Integrin associated protein, IAP
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (1μg)/test