Please login to your online account to display your discounted pricing

CD47 Mouse anti-Human, Clone: B6H12, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$143.55 - $520.55

Specifications

Antigen CD47
Clone B6H12
Host Species Mouse
Gene Alias Integrin associated protein, IAP
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014326

 
life technologies
LS16047981
50μg Each for $143.55

5014327

 
life technologies
LS16047985
500μg Each for $520.55
Description & Specifications

Specifications

Antigen CD47
Clone B6H12
Host Species Mouse
Gene Alias Integrin associated protein, IAP
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The monoclonal antibody B6H12 reacts to CD47 also known as integrin-associated protein (IAP), and neurophilin. CD47 is a glycosylated five transmembrane protein with a small alternatively spliced cytoplasmic domain. CD47 is involved in adhesion through interactions with SIRP (signal regulator protein) and is non-covalently associated with β3 integrins CD51/CD61 and CD41/CD61. Furthermore this interaction can mediate bi-directional signaling to modify neural synaptic activity and regulate the phagocytic activities of macrophages. CD47 is the receptor for thrombospondin. T cell expression of CD47 can mediate activation or apoptosis (in the presence of high levels of thrombospondin). Recently stimulation of CD47 by monoclonal antibody has been shown to induce CD4+CD25- suppressive activity also increasing expression of Foxp3. Expression is found in the majority of hematopoietic cells including T and B cells, monocytes, platelets and erythrocytes (as part of the Rh complex). Expression is also found in non-hematopoietic cells.

This antibody has been reported to have neutralizing activity.