missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD47 Mouse anti-Human, Clone: B6H12, eBioscience™

Mouse Monoclonal Antibody

$91.08 - $143.88

Specifications

Antigen CD47
Clone B6H12
Host Species Murine
Gene Alias Integrin associated protein, IAP
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012616 Life Technologies
LS14047980
25μg Each for $91.08
5012617 Life Technologies
LS14047982
100μg Each for $143.88
Specifications

Description

The monoclonal antibody B6H12 reacts to CD47 also known as integrin-associated protein (IAP), and neurophilin. CD47 is a glycosylated five transmembrane protein with a small alternatively spliced cytoplasmic domain. CD47 is involved in adhesion through interactions with SIRP (signal regulator protein) and is non-covalently associated with β3 integrins CD51/CD61 and CD41/CD61. Furthermore this interaction can mediate bi-directional signaling to modify neural synaptic activity and regulate the phagocytic activities of macrophages. CD47 is the receptor for thrombospondin. T cell expression of CD47 can mediate activation or apoptosis (in the presence of high levels of thrombospondin). Recently stimulation of CD47 by monoclonal antibody has been shown to induce CD4+CD25- suppressive activity also increasing expression of Foxp3. Expression is found in the majority of hematopoietic cells including T and B cells, monocytes, platelets and erythrocytes (as part of the Rh complex). Expression is also found in non-hematopoietic cells.

This antibody has been reported to have neutralizing activity.
Description & Specifications

Specifications

CD47
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
B6H12
Integrin associated protein, IAP
Flow Cytometry,Immunohistochemistry (Formalin/Paraffin),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL