Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD45RO (T-Cell Marker) Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD45RO (T-Cell Marker) Ab-1, Mouse Monoclonal Antibody.

$231.43 - $628.55

Specifications

Antigen CD45RO (T-Cell Marker) Ab-1
Clone UCHL-1
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS112P

 
thermo scientific™ lab vision™
MS112P
Purified Ab with BSA and Azide 1mL Each for $578.98

MS112P0

 
thermo scientific™ lab vision™
MS112P0
Purified Ab with BSA and Azide 100µL Each for $231.43

MS112P1

 
thermo scientific™ lab vision™
MS112P1
Purified Ab with BSA and Azide 500µL Each for $389.65

MS112P1ABX

 
thermo scientific™ lab vision™
MS112P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $492.18

MS112PABX

 
thermo scientific™ lab vision™
MS112PABX
Ab without BSA/Azide (Frozen) 200µL Each for $628.55

MS112R7

 
thermo scientific™ lab vision™
MS112R7
Ready-to-use for IHC (UVLP) 7mL Each for $334.33
Description & Specifications

Specifications

Antigen CD45RO (T-Cell Marker) Ab-1
Clone UCHL-1
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Mouse
Regulatory Status IVD
Research Discipline Oncogenesis
Type Primary
Immunogen Interleukin-2-dependent human T lymphocytes

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD45RO (T-Cell Marker) Ab-1, Mouse Monoclonal Antibody.

CD45RO (an isoform of leukocyte common antigen) is expressed on most thymocytes, monocytes, macrophages and granulocytes. It is not expressed on normal B lymphocytes. Intrathymic T cell maturation progresses from CD45RA-/CD45RO+ to CD45RA+/CD45RO-; while peripheral T cell maturation progresses from CD45RA+/CD45RO- to CD45RA-/CD45RO+.

Host Species: Mouse

Clone: UCHL-1

Isotype: IgG2a/κ

Species Reactivity: Human, Rhesus Monkey and Chimpanzee. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: Interleukin-2-dependent human T lymphocytes

Molecular Weight: 180 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry