Please login to your online account to display your discounted pricing

CD45RO Mouse anti-Human, PE-Cy7, Clone: UCHL1, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $456.75

Specifications

Antigen CD45RO
Clone UCHL1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015519

 
life technologies
LS25045741
25 Tests Each for $216.05

5015520

 
life technologies
LS25045742
100 Tests Each for $456.75
Description & Specifications

Specifications

Antigen CD45RO
Clone UCHL1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The UCHL1 monoclonal antibody reacts with human CD45RO, a 180kDa isoform of CD45. CD45RO is expressed by most thymocytes, activated memory T cells, granulocytes and monocytes. CD22 is a ligand for CD45RO. Expression of CD45RO and CD45RA is used commonly to discriminate subsets of peripheral T cells.