missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45RB Rat anti-Mouse, FITC, Clone: C363.16A, eBioscience™

Rat Monoclonal Antibody

$71.05 - $390.05

Specifications

Antigen CD45RB
Clone C363.16A
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122064 Life Technologies
LS11045585
500μg Each for $390.05
509585 Life Technologies
LS11045582
100μg Each for $129.05
509584 Life Technologies
LS11045581
50μg Each for $71.05
Specifications

Description

The C363.16A monoclonal antibody reacts with the exon B-dependent isoform of mouse CD45. This CD45 isoform is expressed by thymocytes, peripheral B cells and a subpopulation of T cells at high density and by dendritic cells and macrophages at low density. CD45 is a protein tyrosine phosphatase and serves an important role in leukocyte signal transduction.
Description & Specifications

Specifications

CD45RB
Rat
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
C363.16A
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer