Please login to your online account to display your discounted pricing

CD45RA Mouse anti-Rat, APC, Clone: OX33, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $332.05

Specifications

Antigen CD45RA
Clone OX33
Host Species Mouse
Gene Alias Leukocyte Common Antigen
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245648

 
life technologies
LS17046280
25μg Each for $143.55

50245649

 
life technologies
LS17046282
100μg Each for $332.05
Description & Specifications

Specifications

Antigen CD45RA
Clone OX33
Host Species Mouse
Gene Alias Leukocyte Common Antigen
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This OX33 monoclonal antibody reacts with rat CD45RA. In rats, this isoform of CD45, which is also known as leukocyte common antigen, is present only on B cells. CD45 is a type I transmembrane protein tyrosine phosphatase that exists as a variety of different isoforms due to alternative splicing.