missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45RA Mouse anti-Human, PE-eFluor 610, Clone: HI100, eBioscience™

Mouse Monoclonal Antibody

$161.04 - $402.60

Specifications

Antigen CD45RA
Clone HI100
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124765 Life Technologies
LS61045841
25 Tests Each for $161.04
501129198 Life Technologies
LS61045842
100 Tests Each for $402.60
Specifications

Description

The HI100 monoclonal antibody reacts with human CD45RA, a 220kDa molecule expressed by subpopulations of CD4+ peripheral T lymphocytes, CD8+ peripheral T lymphocytes, and B cells. The CD45RA+ T cell populations are mainly naive/virgin allowing the use of HI100 mAb as a phenotypic marker to discriminate T cell subsets.
Description & Specifications

Specifications

CD45RA
Murine
Flow Cytometry
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.03μg)/test
HI100
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer