Please login to your online account to display your discounted pricing

CD45RA Mouse anti-Human, PE-Cy5, Clone: HI100, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $375.55

Specifications

Antigen CD45RA
Clone HI100
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014064

 
life technologies
LS15045842
100 tests Each for $375.55

5014063

 
life technologies
LS15045841
25 tests Each for $172.55
Description & Specifications

Specifications

Antigen CD45RA
Clone HI100
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The HI100 monoclonal antibody reacts with human CD45RA, a 220kDa molecule expressed by subpopulations of CD4+ peripheral T lymphocytes, CD8+ peripheral T lymphocytes, and B cells. The CD45RA+ T cell populations are mainly naive/virgin allowing the use of HI100 mAb as a phenotypic marker to discriminate T cell subsets.