missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45RA Mouse anti-Human, FITC, Clone: HI100, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD45RA
Clone HI100
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122065 Life Technologies
LS11045841
25 Tests Each for $114.55
501122066 Life Technologies
LS11045842
100 Tests Each for $259.55
Specifications

Description

The HI100 monoclonal antibody reacts with human CD45RA, a 220kDa molecule expressed by subpopulations of CD4+ peripheral T lymphocytes, CD8+ peripheral T lymphocytes, and B cells. The CD45RA+ T cell populations are mainly naïve/virgin allowing the use of HI100 mAb as a phenotypic marker to discriminate T cell subsets.
Description & Specifications

Specifications

CD45RA
Murine
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (1μg)/test
HI100
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer