missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD45RA Mouse anti-Human, Clone: HI100, eBioscience™

Mouse Monoclonal Antibody

$77.88 - $143.88

Specifications

Antigen CD45RA
Clone HI100
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry,Immunohistochemistry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012607 Life Technologies
LS14045880
25μg Each for $77.88
5012608 Life Technologies
LS14045882
100μg Each for $143.88
Specifications

Description

The HI100 monoclonal antibody reacts with human CD45RA, a 220kDa molecule expressed by subpopulations of CD4+ peripheral T lymphocytes, CD8+ peripheral T lymphocytes, and B cells. The CD45RA+ T cell populations are mainly naive/virgin allowing the use of HI100 mAb as a phenotypic marker to discriminate T cell subsets.
Description & Specifications

Specifications

CD45RA
Murine
Flow Cytometry,Immunohistochemistry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
HI100
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer